Learn More
Medchemexpress LLC UNC2025 hydrochloride | 2070015-17-5 | 100.0% | 513.12 | C28H41ClN6O | 10MM 1ML
SDP
Catalog No. 5000236412
Encompass_Preferred
UNC2025 hydrochloride is the hydrochloride salt of UNC2025, a potent, ATP-competitive, orally active dual MERTK/FLT3 inhibitor used in preclinical research on acute leukemia and receptor tyrosine kinase signaling. Supplied as a 10 mM solution in DMSO (1 mL), it exhibits sub-nanomolar inhibition of MERTK and FLT3 and demonstrates favorable pharmacokinetic properties for in vitro and in vivo studies.
  • Potent MERTK and FLT3 inhibition at sub-nanomolar concentrations.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • High purity for reproducible experimental results.
  • Light yellow to yellow solid appearance when provided as a dry compound.
  • Store sealed and protect from moisture; in solution store at -80°C for long-term stability.
Catalog No. 50-002-36412 Supplier Medchemexpress LLC Supplier No. HY12344A10MM 1ML
UNC2025 hydrochloride is the hydrochloride salt of UNC2025, a potent, ATP-competitive, orally active dual MERTK/FLT3 inhibitor used in preclinical research on acute leukemia and receptor tyrosine kinase signaling. Supplied as a 10 mM solution in DMSO (1 mL), it exhibits sub-nanomolar inhibition of MERTK and FLT3 and demonstrates favorable pharmacokinetic properties for in vitro and in vivo studies.
  • Potent MERTK and FLT3 inhibition at sub-nanomolar concentrations.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • High purity for reproducible experimental results.
  • Light yellow to yellow solid appearance when provided as a dry compound.
  • Store sealed and protect from moisture; in solution store at -80°C for long-term stability.