Learn More
Medchemexpress LLC UNC2025 hydrochloride | 2070015-17-5 | 100.0% | 513.12 | C28H41ClN6O | 25MG
SDP
Catalog No. 5000237449
Encompass_Preferred
UNC2025 hydrochloride is the hydrochloride salt of UNC2025, a potent, ATP-competitive, orally active dual Mer (MERTK) and FLT3 kinase inhibitor used in preclinical oncology research. It exhibits sub-nanomolar potency and favorable pharmacokinetic properties, and is supplied as a light yellow solid for laboratory use.
  • Potent Mer and FLT3 inhibition (IC50 0.74 nM and 0.8 nM).
  • High reported purity (99.96%).
  • Solid form with light yellow to yellow appearance.
  • Demonstrated >45-fold selectivity for MERTK versus Axl.
  • Available in multiple pack sizes and solution aliquots for flexible dosing.
  • Useful as a tool compound in preclinical oncology models, including acute leukemia research.
Catalog No. 50-002-37449 Supplier Medchemexpress LLC Supplier No. HY12344A25MG
UNC2025 hydrochloride is the hydrochloride salt of UNC2025, a potent, ATP-competitive, orally active dual Mer (MERTK) and FLT3 kinase inhibitor used in preclinical oncology research. It exhibits sub-nanomolar potency and favorable pharmacokinetic properties, and is supplied as a light yellow solid for laboratory use.
  • Potent Mer and FLT3 inhibition (IC50 0.74 nM and 0.8 nM).
  • High reported purity (99.96%).
  • Solid form with light yellow to yellow appearance.
  • Demonstrated >45-fold selectivity for MERTK versus Axl.
  • Available in multiple pack sizes and solution aliquots for flexible dosing.
  • Useful as a tool compound in preclinical oncology models, including acute leukemia research.