Learn More
Medchemexpress LLC UNC2025 hydrochloride | 2070015-17-5 | MFCD28963885 | 100.0% | 513.12 g/mol | C28H41ClN6O | 100MG
SDP
Catalog No. 5000238726
Encompass_Preferred
UNC2025 hydrochloride is an ATP-competitive, orally active inhibitor of Mer and FLT3 kinases used in preclinical leukemia research and kinase signaling studies. It demonstrates potent inhibition, selectivity for MERTK, and stability suitable for laboratory workflows.
  • Sub-nanomolar potency against Mer and FLT3.
  • Greater than 45-fold selectivity for MERTK versus Axl.
  • High purity solid form suitable for biochemical and cellular assays.
  • Available in multiple quantities including 100 mg for in vivo or dosing studies.
  • Stable when stored sealed at 4°C; in solution store at -80°C for long term.
Catalog No. 50-002-38726 Supplier Medchemexpress LLC Supplier No. HY12344A100MG
UNC2025 hydrochloride is an ATP-competitive, orally active inhibitor of Mer and FLT3 kinases used in preclinical leukemia research and kinase signaling studies. It demonstrates potent inhibition, selectivity for MERTK, and stability suitable for laboratory workflows.
  • Sub-nanomolar potency against Mer and FLT3.
  • Greater than 45-fold selectivity for MERTK versus Axl.
  • High purity solid form suitable for biochemical and cellular assays.
  • Available in multiple quantities including 100 mg for in vivo or dosing studies.
  • Stable when stored sealed at 4°C; in solution store at -80°C for long term.