Learn More
Cayman Chemical UNC2170maleate 10mg
SDP
Catalog No. 502968068
Encompass_Preferred
A micromolar ligand of 53BP1 that binds to a pocket formed by the tandem tudor domains on a 53BP1 homodimer; functions as a 53BP1 antagonist in cellular lysates
Catalog No. 50-296-8068 Supplier Cayman Chemical Supplier No. 1737410 MG
A micromolar ligand of 53BP1 that binds to a pocket formed by the tandem tudor domains on a 53BP1 homodimer; functions as a 53BP1 antagonist in cellular lysates