Learn More
Medchemexpress LLC HY-125510 5mg Medchemexpress, UNC2541 CAS:1612782-86-1 Purity:>98%
SDP
Catalog No. 501932607
Encompass_Preferred
Medchemexpress, HY-125510 5mg UNC2541 CAS:1612782-86-1 UNC2541 is a potent and Mer tyrosine kinase ( MerTK )-specific inhibitor, binds in the MerTK ATP pocket, with an IC 50 of 4.4 nM, more selective over Axl, Tyro3 and Flt3. UNC2541 inhibits phosphorylated MerTK (pMerTK; EC 50 , 510 nM) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2607 Supplier Medchemexpress LLC Supplier No. HY1255105MG
Medchemexpress, HY-125510 5mg UNC2541 CAS:1612782-86-1 UNC2541 is a potent and Mer tyrosine kinase ( MerTK )-specific inhibitor, binds in the MerTK ATP pocket, with an IC 50 of 4.4 nM, more selective over Axl, Tyro3 and Flt3. UNC2541 inhibits phosphorylated MerTK (pMerTK; EC 50 , 510 nM) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.