Learn More
Medchemexpress LLC HY-110150 5mg Medchemexpress, UNC3230 CAS:1031602-63-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1101505MG
Encompass_Preferred
Medchemexpress, HY-110150 5mg UNC3230 CAS:1031602-63-7 UNC3230 is a potent, selective and ATP-competitive PIP5K1C inhibitor with an IC50 of ~41 nM. UNC3230 also inhibits PIP4K2C and does not inhibit any of the other lipid kinases that regulate phosphoinositide levels. UNC3230 has antinociceptive and anticancer effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9677