Learn More
Aviva Systems Biology Corp UNC45A Antibody - N-terminal region (ARP56261_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56261_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UNC45A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human UNC45A. Alternate names: FLJ10043, GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-133