Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UNC4976 trifluoroacetate | 98.0% | 961.12 g/mol | C49H71F3N6O10 | 5 MG
SDP

Catalog No. 5000264067
Encompass_Preferred

UNC4976 TFA is the trifluoroacetate salt of UNC4976, a peptidomimetic positive allosteric modulator (PAM) of the CBX7 chromodomain that modulates chromodomain-nucleic acid interactions. It is supplied as a purified research compound intended for in vitro studies in epigenetics and chromatin biology.

  • Trifluoroacetate salt form for improved handling and stability.
  • High purity suitable for research applications (98.0%).
  • Molecular formula C49H71F3N6O10 and molecular weight 961.12 g/mol.
  • Available in small pack sizes for screening and assay development.
  • Suitable for use in studies of chromodomain function and epigenetic modulation.
  • Provided with documentation specifying identity and purity.

Catalog No. 50-002-64067 Supplier Medchemexpress LLC Supplier No. HY126327A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UNC4976 TFA is the trifluoroacetate salt of UNC4976, a peptidomimetic positive allosteric modulator (PAM) of the CBX7 chromodomain that modulates chromodomain-nucleic acid interactions. It is supplied as a purified research compound intended for in vitro studies in epigenetics and chromatin biology.

  • Trifluoroacetate salt form for improved handling and stability.
  • High purity suitable for research applications (98.0%).
  • Molecular formula C49H71F3N6O10 and molecular weight 961.12 g/mol.
  • Available in small pack sizes for screening and assay development.
  • Suitable for use in studies of chromodomain function and epigenetic modulation.
  • Provided with documentation specifying identity and purity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.