Learn More
Enzo Life Sciences UNC5B monoclonal antibody (Guido-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804846C100
Encompass_Preferred
Clone: Guido-1. Isotype: IgG2b. Immunogen: Recombinant human UNC5B (aa 28-363). Source: Purified from ascites. Species reactivity: Human, Mouse. Applications: Flow Cytometry. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9999