Learn More
Aviva Systems Biology Corp UNC84B Antibody - C-terminal region (ARP47148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UNC84B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human UNC84B. Alternate names: RP3-508I15.15, KIAA0668, MGC133055, MGC133056, SUN2, UNC84BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53507