Learn More
Aviva Systems Biology Corp UNC93B1 Antibody - N-terminal region (ARP49979_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49979_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against UNC93B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UNC93B1. Alternate names: MGC126617, UNC93, UNC93B, IIAE1, Unc-93B1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53611