Learn More
Medchemexpress LLC UNC9994 hydrochloride | 2108826-33-9 | 99.5% | 457.84 | 10 MG
SDP
Catalog No. 5000343388
Encompass_Preferred
UNC9994 hydrochloride is a functionally selective, β-arrestin-biased dopamine D2 receptor (D2R) agonist. It selectively activates β-arrestin recruitment and signaling, demonstrating antipsychotic-like activity. It also acts as an antagonist of Gi-regulated cAMP production and a partial agonist for D2R/β-arrestin-2 interactions.
  • Selectively activates β-arrestin recruitment and signaling.
  • Antagonist of Gi-regulated cAMP production.
  • Partial agonist for D2R/β-arrestin-2 interactions.
  • Shows antipsychotic-like activity.
  • Induces D2-mediated β-arrestin-2 translocation with EC50s of 6.1 nM and 448 nM.
  • Acts as an antagonist at 5HT2A and 5HT2B receptors.
  • Acts as an agonist at 5HT2C and 5HT1A receptors.
Catalog No. 50-003-43388 Supplier Medchemexpress LLC Supplier No. HY111385100MG
UNC9994 hydrochloride is a functionally selective, β-arrestin-biased dopamine D2 receptor (D2R) agonist. It selectively activates β-arrestin recruitment and signaling, demonstrating antipsychotic-like activity. It also acts as an antagonist of Gi-regulated cAMP production and a partial agonist for D2R/β-arrestin-2 interactions.
  • Selectively activates β-arrestin recruitment and signaling.
  • Antagonist of Gi-regulated cAMP production.
  • Partial agonist for D2R/β-arrestin-2 interactions.
  • Shows antipsychotic-like activity.
  • Induces D2-mediated β-arrestin-2 translocation with EC50s of 6.1 nM and 448 nM.
  • Acts as an antagonist at 5HT2A and 5HT2B receptors.
  • Acts as an agonist at 5HT2C and 5HT1A receptors.