Learn More
Medchemexpress LLC UNC9994 hydrochloride | 2108826-33-9 | 99.5% | 457.84 | 100 MG
SDP
Catalog No. 5000346333
Encompass_Preferred
UNC9994 hydrochloride is a functionally selective, β-arrestin-biased dopamine D2 receptor (D2R) agonist that selectively activates β-arrestin recruitment and signaling. It shows a binding affinity with a Ki of 79 nM for D2R. UNC9994 hydrochloride is also an antagonist of Gi-regulated cAMP production and a partial agonist for D2R/β-arrestin-2 interactions. It demonstrates antipsychotic-like activity. It is for research use only.
  • Functions as a β-arrestin-biased dopamine D2 receptor agonist.
  • Selectively activates β-arrestin recruitment and signaling.
  • Exhibits a binding affinity (Ki) of 79 nM for D2R.
  • Acts as an antagonist of Gi-regulated cAMP production.
  • Serves as a partial agonist for D2R/β-arrestin-2 interactions.
  • Demonstrates antipsychotic-like activity.
Catalog No. 50-003-46333 Supplier Medchemexpress LLC Supplier No. HY11138550MG
UNC9994 hydrochloride is a functionally selective, β-arrestin-biased dopamine D2 receptor (D2R) agonist that selectively activates β-arrestin recruitment and signaling. It shows a binding affinity with a Ki of 79 nM for D2R. UNC9994 hydrochloride is also an antagonist of Gi-regulated cAMP production and a partial agonist for D2R/β-arrestin-2 interactions. It demonstrates antipsychotic-like activity. It is for research use only.
  • Functions as a β-arrestin-biased dopamine D2 receptor agonist.
  • Selectively activates β-arrestin recruitment and signaling.
  • Exhibits a binding affinity (Ki) of 79 nM for D2R.
  • Acts as an antagonist of Gi-regulated cAMP production.
  • Serves as a partial agonist for D2R/β-arrestin-2 interactions.
  • Demonstrates antipsychotic-like activity.