Learn More
Medchemexpress LLC UNC9994 hydrochloride | 2108826-33-9 | 99.5% | 457.84 | 25 MG
SDP
Catalog No. 5000346191
Encompass_Preferred
UNC9994 hydrochloride is a functionally selective, β-arrestin-biased dopamine D2 receptor (D2R) agonist that selectively activates β-arrestin recruitment and signaling. It exhibits a binding affinity with a Ki of 79 nM for D2R. This compound also acts as an antagonist of Gi-regulated cAMP production and a partial agonist for D2R/β-arrestin-2 interactions. UNC9994 hydrochloride demonstrates antipsychotic-like activity. For research use only.
  • Selectively activates β-arrestin recruitment and signaling
  • Exhibits binding affinity with a Ki of 79 nM for D2R
  • Acts as an antagonist of Gi-regulated cAMP production
  • Functions as a partial agonist for D2R/β-arrestin-2 interactions
  • Demonstrates antipsychotic-like activity
Catalog No. 50-003-46191 Supplier Medchemexpress LLC Supplier No. 25MGHY111385
UNC9994 hydrochloride is a functionally selective, β-arrestin-biased dopamine D2 receptor (D2R) agonist that selectively activates β-arrestin recruitment and signaling. It exhibits a binding affinity with a Ki of 79 nM for D2R. This compound also acts as an antagonist of Gi-regulated cAMP production and a partial agonist for D2R/β-arrestin-2 interactions. UNC9994 hydrochloride demonstrates antipsychotic-like activity. For research use only.
  • Selectively activates β-arrestin recruitment and signaling
  • Exhibits binding affinity with a Ki of 79 nM for D2R
  • Acts as an antagonist of Gi-regulated cAMP production
  • Functions as a partial agonist for D2R/β-arrestin-2 interactions
  • Demonstrates antipsychotic-like activity