Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UNC9994 hydrochloride | 2108826-33-9 | 99.5% | 457.84 | 50 MG
SDP

Catalog No. 5000343372
Encompass_Preferred

UNC9994 hydrochloride | 2108826-33-9 | 99.5% | 457.84 | 50 MG

Catalog No. 50-003-43372 Supplier Medchemexpress LLC Supplier No. HY11138510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UNC9994 hydrochloride | 2108826-33-9 | 99.5% | 457.84 | 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.