Learn More
Ibidi USA µ-Dish 35 mm Quad, Uncoated - Uncoated Polymer Coverslip
SDP
Catalog No. NC1431907
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Dish 35mm Quad allows you to perform high resolution microscopy in a 35 mm Petri-dish with a 4 well subdivision. The Ph+ structure in the center provides excellent phase contrast without a distracting meniscus. The lid can be closed to reduce evaporation during long term experiments. #1.5 polymer coverslip, hydrophobic, sterilized, 60 pcs./box
This item is not returnable. View return policy
Catalog No. NC1431907 Supplier Ibidi USA Supplier No. 80411
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Dish 35mm Quad allows you to perform high resolution microscopy in a 35 mm Petri-dish with a 4 well subdivision. The Ph+ structure in the center provides excellent phase contrast without a distracting meniscus. The lid can be closed to reduce evaporation during long term experiments. #1.5 polymer coverslip, hydrophobic, sterilized, 60 pcs./box