Learn More
Vector Laboratories Aleuria aurantia lectin (AAL) Unconjugated 2 mg
SDP
Catalog No. NC1409924
Encompass
Aleuria aurantia lectin (AAL) binds preferentially to fucose linked (α -1,6) to N-acetylglucosamine or to fucose linked (α -1,3) to N-acetyllactosamine-related structures, unlike Ulex europaeus and Lotus tetragonolobus lectins, which prefer (α -1,2)-linked fucose residues. AAL also reversibly binds fucose that is attached to nucleic acids.
Catalog No. NC1409924 Supplier Vector Laboratories Supplier No. L13902
Aleuria aurantia lectin (AAL) binds preferentially to fucose linked (α -1,6) to N-acetylglucosamine or to fucose linked (α -1,3) to N-acetyllactosamine-related structures, unlike Ulex europaeus and Lotus tetragonolobus lectins, which prefer (α -1,2)-linked fucose residues. AAL also reversibly binds fucose that is attached to nucleic acids.