Learn More
Jackson Immuno Research Labs AffiniPure Donkey Anti-Human IgG (H+L), 1mg
SDP
Catalog No. NC9873589
Encompass_Preferred
AffiniPure Donkey Anti-Human IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Ms,Rb,Rat,Shp Sr Prot)
Catalog No. NC9873589 Supplier Jackson Immuno Research Labs Supplier No. 709005149
AffiniPure Donkey Anti-Human IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Ms,Rb,Rat,Shp Sr Prot)