Learn More
ACROBiosystems Unconjugated Human HLA-A*24:02&B2M&EBV BRLF1 (TYPVLEEMF) Complex Protein (Monomer, MALS verified)
Catalog No. 502719456
Encompass_Preferred
Unconjugated Human HLA-A*24:02&B2M&EBV BRLF1 (TYPVLEEMF) Complex Protein (HLF-H52E5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*24:02) & Ile 21 - Met 119 (B2M) & TYPVLEEMF peptide (Accession # AAA59600.1 (HLA-A*24:02) & P61769 (B2M) & TYPVLEEMF).
Catalog No. 50-271-9456 Supplier ACROBiosystems Supplier No. HLFH52E525UG
Unconjugated Human HLA-A*24:02&B2M&EBV BRLF1 (TYPVLEEMF) Complex Protein (HLF-H52E5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*24:02) & Ile 21 - Met 119 (B2M) & TYPVLEEMF peptide (Accession # AAA59600.1 (HLA-A*24:02) & P61769 (B2M) & TYPVLEEMF).