Learn More
Vector Laboratories Sambucus Nigra Lectin (SNA, EBL) Unconjugated 5 mg
SDP
Catalog No. NC9061539
Encompass
Sambucus nigra agglutinin (SNA) is a lectin isolated from elderberry bark that binds preferentially to sialic acid attached to terminal galactose in α-2,6 and to a lesser degree, α-2,3 linkage. Binding is also inhibited to some extent by lactose or galactose. This lectin does not appear to bind sialic acid linked to N-acetylgalactosamine. SNA has been reported to inhibit cell-free protein synthesis.
Catalog No. NC9061539 Supplier Vector Laboratories Supplier No. L13005
Sambucus nigra agglutinin (SNA) is a lectin isolated from elderberry bark that binds preferentially to sialic acid attached to terminal galactose in α-2,6 and to a lesser degree, α-2,3 linkage. Binding is also inhibited to some extent by lactose or galactose. This lectin does not appear to bind sialic acid linked to N-acetylgalactosamine. SNA has been reported to inhibit cell-free protein synthesis.