Learn More
Aviva Systems Biology Corp UNCX Antibody - C-terminal region (ARP47546_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47546_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UNCX. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human UNCX. Alternate names: UNCX4.1ApplicationIHC, WB. Species reactivity: Human, Rat, Zebrafish
Catalog No. 50-014-59583