Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecanal 97.0+%, TCI America™
SDP

Catalog No. U0009250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U0009250ML 250 mL
U000925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U0009250ML Supplier TCI America Supplier No. U0009250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-44-7
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00007016
InChI Key KMPQYAYAQWNLME-UHFFFAOYSA-N
Synonym undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde
PubChem CID 8186
ChEBI CHEBI:46202
IUPAC Name undecanal
SMILES CCCCCCCCCCC=O

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 80°C
Quantity 250 mL
Formula Weight 170.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecanal
EINECSNumber (2)-0494
RTECSNumber YQ1500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.