Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019912 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9912 5 mL
11-101-8403 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9912 Supplier MilliporeSigma Supelco Supplier No. 940005ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Undecane is obtained from the refining of petroleum. It is a component of gasoline and is generally used in organic synthesis, petroleum research and as a distillation chaser.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 1120-21-4
Vapor Pressure <0.4 mmHg (20°C)
Vapor Density 5.4 (vs. air)
Density 0.74 g/mL (at 25°C)
Flash Point 62°C
Physical Form Neat
Linear Formula CH3(CH2)9 CH3
MDL Number MFCD00008959
Melting Point -26°C
Boiling Point 196°C
Synonym n-Undecane; Hendecane
Type Analytical Standard
Quantity 5 mL
Formula Weight 156.31
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.