Learn More
Medchemexpress LLC Undecanedioic acid | 1852-04-6 | MFCD00004444 | ≥98.0% | 216.28 g/mol | C11H20O4 | 1 ML
SDP
Catalog No. 5000304032
Encompass_Preferred
Undecanedioic acid is an endogenous dicarboxylic acid associated with extracellular matrix macromolecules, particularly elastin. It is supplied for research and analytical use as a solid or as a 10 mM solution in DMSO (purity ≥98%, molecular weight 216.28 g/mol).
  • Endogenous dicarboxylic acid associated with elastin.
  • Available as a solid or 10 mM solution in DMSO.
  • High purity suitable for analytical and research applications.
  • Molecular formula C11H20O4 and molecular weight 216.28 g/mol.
  • Store and handle according to the SDS; keep sealed and away from moisture.
Catalog No. 50-003-04032 Supplier Medchemexpress LLC Supplier No. HYW01412510MM/1ML
Add to Cart
Add to Cart
Undecanedioic acid is an endogenous dicarboxylic acid associated with extracellular matrix macromolecules, particularly elastin. It is supplied for research and analytical use as a solid or as a 10 mM solution in DMSO (purity ≥98%, molecular weight 216.28 g/mol).
  • Endogenous dicarboxylic acid associated with elastin.
  • Available as a solid or 10 mM solution in DMSO.
  • High purity suitable for analytical and research applications.
  • Molecular formula C11H20O4 and molecular weight 216.28 g/mol.
  • Store and handle according to the SDS; keep sealed and away from moisture.