Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Undecanedioic acid | 1852-04-6 | MFCD00004444 | 98.0% | 216.28 g/mol | C11H20O4 | 100 G
SDP

Catalog No. 5000302096
Encompass_Preferred

Undecanedioic acid is a linear dicarboxylic acid used as an analytical reference standard and an endogenous metabolite in biochemical and analytical research. Supplied as a solid, it is suitable for analytical method development, metabolite identification, and materials research.

  • High purity: 98.0%.
  • Chemical formula C11H20O4 and molecular weight 216.28 g/mol.
  • Available in multiple pack sizes including 100 g.
  • Soluble in DMSO; solutions are available.
  • Suitable for analytical and metabolite reference applications.

Catalog No. 50-003-02096 Supplier Medchemexpress LLC Supplier No. HYW014125100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Undecanedioic acid is a linear dicarboxylic acid used as an analytical reference standard and an endogenous metabolite in biochemical and analytical research. Supplied as a solid, it is suitable for analytical method development, metabolite identification, and materials research.

  • High purity: 98.0%.
  • Chemical formula C11H20O4 and molecular weight 216.28 g/mol.
  • Available in multiple pack sizes including 100 g.
  • Soluble in DMSO; solutions are available.
  • Suitable for analytical and metabolite reference applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.