Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Undecanedioic acid | 1852-04-6 | MFCD00004444 | ≥98.0% | 216.28 g/mol | C11H20O4 | 25 G
SDP

Catalog No. 5000303544
Encompass_Preferred

Undecanedioic acid is an endogenous medium-chain dicarboxylic acid supplied as a solid analytical reagent for biochemical and analytical applications. It is associated with intercellular matrix macromolecules, particularly elastin, and is commonly used as a reference standard or research reagent in metabolic and structural studies.

  • Endogenous dicarboxylic acid associated with elastin.
  • CAS number 1852-04-6.
  • Chemical formula C11H20O4; molecular weight 216.28 g/mol.
  • Purity ≥98.0%.
  • White to off-white solid appearance.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance may be required.
  • Store sealed away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
  • Available in multiple laboratory-scale package sizes including 25 g.

Catalog No. 50-003-03544 Supplier Medchemexpress LLC Supplier No. 25GHYW014125
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Undecanedioic acid is an endogenous medium-chain dicarboxylic acid supplied as a solid analytical reagent for biochemical and analytical applications. It is associated with intercellular matrix macromolecules, particularly elastin, and is commonly used as a reference standard or research reagent in metabolic and structural studies.

  • Endogenous dicarboxylic acid associated with elastin.
  • CAS number 1852-04-6.
  • Chemical formula C11H20O4; molecular weight 216.28 g/mol.
  • Purity ≥98.0%.
  • White to off-white solid appearance.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance may be required.
  • Store sealed away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
  • Available in multiple laboratory-scale package sizes including 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.