Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Undecanoic acid, 11-amino- | 2432-99-7 | MFCD00008150 | ≥97.0% | 201.31 g/mol | C11H23NO2 | 10 G
SDP

Catalog No. 5000303897
Encompass_Preferred

11-Aminoundecanoic acid (C11H23NO2, MW 201.31 g/mol, CAS 2432-99-7) is a medium-chain amino acid used as an alkyl-chain linker in targeted protein degradation chemistry and as a versatile building block in medicinal chemistry research.

  • Alkyl-chain linker suitable for PROTAC construction.
  • Contains a terminal primary amine and carboxylic acid for orthogonal coupling reactions.
  • Molecular weight 201.31 g/mol and formula C11H23NO2.
  • Purity ≥97.0% by NMR according to the certificate of analysis.
  • Storage: stable at room temperature for ambient storage; in solvent, store at -80°C or -20°C for extended periods.

Catalog No. 50-003-03897 Supplier Medchemexpress LLC Supplier No. HYW01483110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

11-Aminoundecanoic acid (C11H23NO2, MW 201.31 g/mol, CAS 2432-99-7) is a medium-chain amino acid used as an alkyl-chain linker in targeted protein degradation chemistry and as a versatile building block in medicinal chemistry research.

  • Alkyl-chain linker suitable for PROTAC construction.
  • Contains a terminal primary amine and carboxylic acid for orthogonal coupling reactions.
  • Molecular weight 201.31 g/mol and formula C11H23NO2.
  • Purity ≥97.0% by NMR according to the certificate of analysis.
  • Storage: stable at room temperature for ambient storage; in solvent, store at -80°C or -20°C for extended periods.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.