Learn More
Medchemexpress LLC Undecanoic acid, 11-azido-, 2,5-dioxo-1-pyrrolidinyl ester | 850080-13-6 | MFCD29041759 | 98.0% | 324.38 g/mol | C15H24N4O4 | 100 MG
SDP
Catalog No. 5000282248
Encompass_Preferred
2,5-Dioxopyrrolidin-1-yl 11-azidoundecanoate is an azide-bearing N-hydroxysuccinimide (NHS) ester supplied as a powder for use in bioconjugation and biochemical assays. It combines an activated carboxyl group for efficient coupling to amines with a terminal azide for click chemistry, enabling versatile linker and probe synthesis in research applications.
  • Activated NHS ester for efficient amide coupling to primary amines
  • Terminal azide functionality for copper-catalyzed and strain-promoted click reactions
  • High purity suitable for biochemical assay reagent applications
  • Supplied as a stable powder for convenient storage and handling
  • Compatible with common solvents used in bioconjugation workflows
Catalog No. 50-002-82248 Supplier Medchemexpress LLC Supplier No. HYW190926100MG
2,5-Dioxopyrrolidin-1-yl 11-azidoundecanoate is an azide-bearing N-hydroxysuccinimide (NHS) ester supplied as a powder for use in bioconjugation and biochemical assays. It combines an activated carboxyl group for efficient coupling to amines with a terminal azide for click chemistry, enabling versatile linker and probe synthesis in research applications.
  • Activated NHS ester for efficient amide coupling to primary amines
  • Terminal azide functionality for copper-catalyzed and strain-promoted click reactions
  • High purity suitable for biochemical assay reagent applications
  • Supplied as a stable powder for convenient storage and handling
  • Compatible with common solvents used in bioconjugation workflows