Learn More
Medchemexpress LLC Undecanoic acid, 11-azido-, 2,5-dioxo-1-pyrrolidinyl ester | 850080-13-6 | MFCD29041759 | >98.0% | 324.38 g/mol | C15H24N4O4 | 25 MG
SDP
Catalog No. 5000283077
Encompass_Preferred
2,5-Dioxo-1-pyrrolidinyl 11-azidoundecanoate is an N-hydroxysuccinimide (NHS) ester of 11-azidoundecanoic acid used as an azide-functional linker for bioconjugation and labeling in biochemical assays. Supplied as a powder, it provides a reactive NHS group for amine coupling and an azide handle for bioorthogonal chemistry.
Catalog No. 50-002-83077 Supplier Medchemexpress LLC Supplier No. HYW19092625MG
2,5-Dioxo-1-pyrrolidinyl 11-azidoundecanoate is an N-hydroxysuccinimide (NHS) ester of 11-azidoundecanoic acid used as an azide-functional linker for bioconjugation and labeling in biochemical assays. Supplied as a powder, it provides a reactive NHS group for amine coupling and an azide handle for bioorthogonal chemistry.