Learn More
Medchemexpress LLC Undecanoic acid | 112-37-8 | 186.29 | 1 ML
SDP
Catalog No. 5000447502
Encompass_Preferred
Undecanoic acid is a monocarboxylic acid known for its antifungal properties. It acts by inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in *T. rubrum*. Additionally, it demonstrates strong antifungal activity by disrupting crucial fungal mechanisms including cell wall synthesis, membrane assembly, mRNA synthesis, lipid metabolism, and pathogenesis, ultimately influencing gene splicing.
  • Monocarboxylic acid
  • Possesses antifungal properties
  • Inhibits exocellular keratinase, lipase, and phospholipid biosynthesis in *T. rubrum*
  • Disrupts fungal cell wall synthesis and membrane assembly
  • Inhibits mRNA synthesis, lipid metabolism, and pathogenesis
  • Influences gene splicing
  • Human endogenous metabolite
Catalog No. 50-004-47502 Supplier Medchemexpress LLC Supplier No. HYW00428210MM/1ML
Undecanoic acid is a monocarboxylic acid known for its antifungal properties. It acts by inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in *T. rubrum*. Additionally, it demonstrates strong antifungal activity by disrupting crucial fungal mechanisms including cell wall synthesis, membrane assembly, mRNA synthesis, lipid metabolism, and pathogenesis, ultimately influencing gene splicing.
  • Monocarboxylic acid
  • Possesses antifungal properties
  • Inhibits exocellular keratinase, lipase, and phospholipid biosynthesis in *T. rubrum*
  • Disrupts fungal cell wall synthesis and membrane assembly
  • Inhibits mRNA synthesis, lipid metabolism, and pathogenesis
  • Influences gene splicing
  • Human endogenous metabolite