Learn More
Medchemexpress LLC Undecanoic acid | 112-37-8 | 186.29 | 50 G
SDP
Catalog No. 5000445767
Encompass_Preferred
Undecanoic acid (Undecanoate) is a monocarboxylic acid that possesses antifungal properties. It functions by inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of various phospholipids in *T. rubrum*. It also demonstrates strong antifungal activity by interfering with critical fungal mechanisms such as cell wall synthesis, membrane assembly, mRNA synthesis, lipid metabolism, and pathogenesis, which can influence gene splicing. As an endogenous metabolite, it shows agonist activity at recombinant human GPR84.
  • Functions as a monocarboxylic acid.
  • Exhibits antifungal properties.
  • Inhibits exocellular keratinase and lipase production.
  • Disrupts fungal cell wall synthesis and membrane assembly.
  • Interferes with fungal mRNA synthesis and lipid metabolism.
  • Influences gene splicing and fungal pathogenesis.
  • Acts as an agonist at recombinant human GPR84.
Catalog No. 50-004-45767 Supplier Medchemexpress LLC Supplier No. HYW00428250G
Undecanoic acid (Undecanoate) is a monocarboxylic acid that possesses antifungal properties. It functions by inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of various phospholipids in *T. rubrum*. It also demonstrates strong antifungal activity by interfering with critical fungal mechanisms such as cell wall synthesis, membrane assembly, mRNA synthesis, lipid metabolism, and pathogenesis, which can influence gene splicing. As an endogenous metabolite, it shows agonist activity at recombinant human GPR84.
  • Functions as a monocarboxylic acid.
  • Exhibits antifungal properties.
  • Inhibits exocellular keratinase and lipase production.
  • Disrupts fungal cell wall synthesis and membrane assembly.
  • Interferes with fungal mRNA synthesis and lipid metabolism.
  • Influences gene splicing and fungal pathogenesis.
  • Acts as an agonist at recombinant human GPR84.