Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Undecanoic acid

Catalog No. 171476100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-147-6100G 100 g
17-147-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-147-6100G Supplier Sigma Aldrich Supplier No. 171476100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Undecanoic acid

Specifications

CAS 112-37-8
Melting Point 28°C to 31°C (lit.)
Boiling Point 228°C (160 mmHg), 248°C to 250°C (lit.)
Molecular Formula C11H22O2
Linear Formula CH3(CH2)9 COOH
MDL Number MFCD00002730
Quantity 100 g
Synonym Hendecanoic acid
Molecular Weight (g/mol) 186.29
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.