Learn More
Medchemexpress LLC Undecanoic acid, biotinyl N-hydroxysuccinimide ester | 887628-40-2 | MFCD05865045 | 99.5% | C25H40N4O6S | 10MG
SDP
Catalog No. 5000259238
Encompass_Preferred
Biotin-C10-NHS Ester is an alkyl/ether-based PROTAC linker that provides an NHS-activated biotinylation reagent for conjugation to primary amines. It is intended for research use in chemical synthesis and bioconjugation workflows, enabling introduction of a biotin affinity tag for purification and detection; follow the manufacturer datasheet and COA for handling and storage instructions.
  • NHS-activated ester for efficient amine coupling.
  • 10-carbon spacer to reduce steric hindrance during binding.
  • Introduces a biotin tag for affinity purification and detection.
  • High purity suitable for synthetic and analytical applications.
  • Supplied with datasheet and certificate of analysis for quality verification.
Catalog No. 50-002-59238 Supplier Medchemexpress LLC Supplier No. HY13592010MG
Biotin-C10-NHS Ester is an alkyl/ether-based PROTAC linker that provides an NHS-activated biotinylation reagent for conjugation to primary amines. It is intended for research use in chemical synthesis and bioconjugation workflows, enabling introduction of a biotin affinity tag for purification and detection; follow the manufacturer datasheet and COA for handling and storage instructions.
  • NHS-activated ester for efficient amine coupling.
  • 10-carbon spacer to reduce steric hindrance during binding.
  • Introduces a biotin tag for affinity purification and detection.
  • High purity suitable for synthetic and analytical applications.
  • Supplied with datasheet and certificate of analysis for quality verification.