Learn More
Medchemexpress LLC Undecanoic acid-d21 | 60658-40-4 | 207.42 | 10 MG
SDP
Catalog No. 5000447514
Encompass_Preferred
Undecanoic acid-d21 is the deuterium labeled Undecanoic acid. It is a monocarboxylic acid with antifungal property, inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. This compound is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-47514 Supplier Medchemexpress LLC Supplier No. HYW004282S10MG
Undecanoic acid-d21 is the deuterium labeled Undecanoic acid. It is a monocarboxylic acid with antifungal property, inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. This compound is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.