Learn More
Medchemexpress LLC Undecanoic acid-d21 | 60658-40-4 | 207.42 | 100 MG
SDP
Catalog No. 5000449133
Encompass_Preferred
Undecanoic acid-d21 is a deuterium-labeled version of undecanoic acid, a monocarboxylic acid with antifungal properties. It functions by inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in *T. rubrum*. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs, making this compound useful for research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits production of exocellular keratinase, lipase, and biosynthesis of phospholipids in *T. rubrum*
  • Suitable for drug development studies due to potential impact on pharmacokinetic and metabolic profiles
Catalog No. 50-004-49133 Supplier Medchemexpress LLC Supplier No. HYW004282S100MG
Undecanoic acid-d21 is a deuterium-labeled version of undecanoic acid, a monocarboxylic acid with antifungal properties. It functions by inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in *T. rubrum*. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs, making this compound useful for research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits production of exocellular keratinase, lipase, and biosynthesis of phospholipids in *T. rubrum*
  • Suitable for drug development studies due to potential impact on pharmacokinetic and metabolic profiles