Learn More
Medchemexpress LLC Undecanoic acid-d21 | 60658-40-4 | 207.42 | 5 MG
SDP
Catalog No. 5000447181
Encompass_Preferred
Undecanoic acid-d21 is a deuterium-labeled monocarboxylic acid known for its antifungal properties. It inhibits the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled undecanoic acid
  • Monocarboxylic acid with antifungal properties
  • Inhibits exocellular keratinase and lipase
  • Inhibits biosynthesis of phospholipids in T. rubrum
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-47181 Supplier Medchemexpress LLC Supplier No. HYW004282S5MG
Undecanoic acid-d21 is a deuterium-labeled monocarboxylic acid known for its antifungal properties. It inhibits the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled undecanoic acid
  • Monocarboxylic acid with antifungal properties
  • Inhibits exocellular keratinase and lipase
  • Inhibits biosynthesis of phospholipids in T. rubrum
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS