Learn More
Medchemexpress LLC Undecanoic acid, ethyl ester | 627-90-7 | 99.7% | 214.35 g/mol | C13H26O2 | 25 G
SDP
Catalog No. 5000304112
Encompass_Preferred
Ethyl undecanoate is the ethyl ester of undecanoic acid supplied as a research reagent and flavor/fragrance intermediate. It is a colorless to light yellow liquid characterized by gas chromatography and proton NMR, commonly used in analytical, formulation, and chemical research applications.
  • High purity (99.68% by GC).
  • Low water content (0.15% by Karl Fischer).
  • Colorless to light yellow liquid appearance.
  • Identity confirmed by proton NMR and gas chromatography.
  • Suitable for flavor, fragrance, and chemical research applications.
  • Available in laboratory-scale packaging (25 g).
Catalog No. 50-003-04112 Supplier Medchemexpress LLC Supplier No. 25GHYW014207
Ethyl undecanoate is the ethyl ester of undecanoic acid supplied as a research reagent and flavor/fragrance intermediate. It is a colorless to light yellow liquid characterized by gas chromatography and proton NMR, commonly used in analytical, formulation, and chemical research applications.
  • High purity (99.68% by GC).
  • Low water content (0.15% by Karl Fischer).
  • Colorless to light yellow liquid appearance.
  • Identity confirmed by proton NMR and gas chromatography.
  • Suitable for flavor, fragrance, and chemical research applications.
  • Available in laboratory-scale packaging (25 g).