Learn More
Medchemexpress LLC Undecanoic acid (Standard) | 112-37-8 | 186.29 | 100 MG
SDP
Catalog No. 5000447993
Encompass_Preferred
Undecanoic acid (Standard) is the analytical standard of Undecanoic acid (HY-W004282) intended for research and analytical applications. It is a monocarboxylic acid with antifungal properties. This compound serves as a reference standard for assays and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Antifungal properties
  • Inhibits production of exocellular keratinase and lipase
  • Inhibits biosynthesis of several phospholipids in T. rubrum
  • Used as an analytical standard
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-47993 Supplier Medchemexpress LLC Supplier No. HYW004282R100MG
Undecanoic acid (Standard) is the analytical standard of Undecanoic acid (HY-W004282) intended for research and analytical applications. It is a monocarboxylic acid with antifungal properties. This compound serves as a reference standard for assays and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Antifungal properties
  • Inhibits production of exocellular keratinase and lipase
  • Inhibits biosynthesis of several phospholipids in T. rubrum
  • Used as an analytical standard
  • Suitable for HPLC, GC, and MS research experiments