Learn More
Medchemexpress LLC Undecanoic acid (standard) | 112-37-8 | 186.29 | 25 MG
SDP
Catalog No. 5000447762
Encompass_Preferred
Undecanoic acid (Undecanoate) is a monocarboxylic acid with antifungal properties. It inhibits the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. This acid exhibits strong antifungal activity by disrupting crucial fungal mechanisms, including cell wall synthesis, membrane assembly, mRNA synthesis, lipid metabolism, and pathogenesis.
  • Monocarboxylic acid with antifungal properties
  • Inhibits exocellular keratinase and lipase production
  • Inhibits phospholipid biosynthesis in T. rubrum
  • Disrupts fungal cell wall synthesis and membrane assembly
  • Impacts fungal mRNA synthesis and lipid metabolism
  • Influences fungal gene splicing and pathogenesis
Catalog No. 50-004-47762 Supplier Medchemexpress LLC Supplier No. 25MGHYW004282R
Undecanoic acid (Undecanoate) is a monocarboxylic acid with antifungal properties. It inhibits the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. This acid exhibits strong antifungal activity by disrupting crucial fungal mechanisms, including cell wall synthesis, membrane assembly, mRNA synthesis, lipid metabolism, and pathogenesis.
  • Monocarboxylic acid with antifungal properties
  • Inhibits exocellular keratinase and lipase production
  • Inhibits phospholipid biosynthesis in T. rubrum
  • Disrupts fungal cell wall synthesis and membrane assembly
  • Impacts fungal mRNA synthesis and lipid metabolism
  • Influences fungal gene splicing and pathogenesis