Learn More
Medchemexpress LLC Undecanoic acid (Standard) | 112-37-8 | 186.29 | 50 MG
SDP
Catalog No. 5000445878
Encompass_Preferred
Undecanoic acid (Standard) is an analytical standard, also known as Undecanoate (Standard) or Hendecanoic acid (Standard), intended for research and analytical applications. This monocarboxylic acid is recognized for its antifungal properties, inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum.
  • Analytical standard for research and analytical applications.
  • Monocarboxylic acid with antifungal properties.
  • Inhibits exocellular keratinase, lipase, and phospholipid biosynthesis in T. rubrum.
  • Used as a reference standard in qualitative, quantitative, and methodological research.
  • Suitable for techniques such as HPLC, GC, and MS.
Catalog No. 50-004-45878 Supplier Medchemexpress LLC Supplier No. HYW004282R50MG
Undecanoic acid (Standard) is an analytical standard, also known as Undecanoate (Standard) or Hendecanoic acid (Standard), intended for research and analytical applications. This monocarboxylic acid is recognized for its antifungal properties, inhibiting the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum.
  • Analytical standard for research and analytical applications.
  • Monocarboxylic acid with antifungal properties.
  • Inhibits exocellular keratinase, lipase, and phospholipid biosynthesis in T. rubrum.
  • Used as a reference standard in qualitative, quantitative, and methodological research.
  • Suitable for techniques such as HPLC, GC, and MS.