Learn More
Medchemexpress LLC Undecanoic-d21 acid | 60658-40-4 | 207.42 | 25 MG
SDP
Catalog No. 5000447852
Encompass_Preferred
Undecanoic acid-d21 is a deuterium labeled form of undecanoic acid, a monocarboxylic acid known for its antifungal properties. It effectively inhibits the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. This compound is suitable for use as a tracer or an internal standard in quantitative analytical methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Exhibits antifungal properties
  • Inhibits exocellular keratinase and lipase
  • Inhibits biosynthesis of phospholipids in T. rubrum
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-47852 Supplier Medchemexpress LLC Supplier No. 25MGHYW004282S
Undecanoic acid-d21 is a deuterium labeled form of undecanoic acid, a monocarboxylic acid known for its antifungal properties. It effectively inhibits the production of exocellular keratinase, lipase, and the biosynthesis of several phospholipids in T. rubrum. This compound is suitable for use as a tracer or an internal standard in quantitative analytical methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Exhibits antifungal properties
  • Inhibits exocellular keratinase and lipase
  • Inhibits biosynthesis of phospholipids in T. rubrum
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS