Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecylcyclohexane 99.0+%, TCI America™
SDP

Catalog No. U003725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U003725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U003725ML Supplier TCI America Supplier No. U003725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54105-66-7
Molecular Formula C17H34
Molecular Weight (g/mol) 238.46
MDL Number MFCD00045505
InChI Key XQQVOBPJWHQXEN-UHFFFAOYSA-N
Synonym 1-Cyclohexylundecane
PubChem CID 40997
IUPAC Name undecylcyclohexane
SMILES CCCCCCCCCCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 165°C
Quantity 25 mL
Formula Weight 238.46
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.