Learn More
Medchemexpress LLC Undecylenate sodium 25g
SDP
Catalog No. 5000205458
Encompass_Preferred
Undecylenate SodiumIt is an organic compound belonging to the class of carboxylates It is formed by the reaction of undecylenic acid and sodium hydroxide Undecylenate SodiumIt has a variety of applications in the cosmetic and personal care industries especially as an antifungal agent in products such as face creams lotions and shampoos In addition it can be used as a preservative in various formulations
Catalog No. 50-002-05458 Supplier Medchemexpress LLC Supplier No. 25GHYW127452
Undecylenate SodiumIt is an organic compound belonging to the class of carboxylates It is formed by the reaction of undecylenic acid and sodium hydroxide Undecylenate SodiumIt has a variety of applications in the cosmetic and personal care industries especially as an antifungal agent in products such as face creams lotions and shampoos In addition it can be used as a preservative in various formulations