Learn More
Sigma Aldrich Fine Chemicals Biosciences Undecylenic acid 96 FG25KG
Catalog No. 502907524
Encompass
Undecylenic acid is also referred to as 10-undecenoic acid. It exhibits anti-bacterial activity against various pathogenic fungi in the pH range of 4.5 to 6.0. It has fungicidal properties.
Catalog No. 50-290-7524 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W32472125KGK
Undecylenic acid is also referred to as 10-undecenoic acid. It exhibits anti-bacterial activity against various pathogenic fungi in the pH range of 4.5 to 6.0. It has fungicidal properties.