Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecylenic Acid, USP, 97-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607643 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
160 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-643 160 kg Amber Glass Bottle
18-607-640 500 mL Amber Glass Bottle
18-607-641 4 L Amber Glass Bottle
18-607-642 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-643 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. UN105160KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

UN105, 112-38-9, C11H20O2

Spectrum™ Chemical Undecylenic Acid, USP is used to treat skin fungal infections such as athlete's foot and jock itch. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 112-38-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
InChI Key FRPZMMHWLSIFAZ-UHFFFAOYSA-N
IUPAC Name undec-10-enoic acid
SMILES OC(=O)CCCCCCCCC=C

Specifications

CAS Max % 1
Color Colorless to pale yellow
Boiling Point 275°C
Ignition Residue 0.0015
Packaging Amber Glass Bottle
Refractive Index 1.447-1.448
Quantity 160 kg
Specific Gravity 0.910-0.913
Percent Purity 97 to 100.5%
Grade USP
Physical Form Liquid
Chemical Name or Material Undecylenic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.