Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ω-Undecylenyl Alcohol 99%, Thermo Scientific™

Catalog No. AC140721000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC140721000 100 g
AC140720250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC140721000 Supplier Thermo Scientific Chemicals Supplier No. 140721000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-43-6
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
InChI Key GIEMHYCMBGELGY-UHFFFAOYSA-N
Synonym 10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcohol
PubChem CID 8185
IUPAC Name undec-10-en-1-ol
SMILES C=CCCCCCCCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.846
Infrared Spectrum Authentic
Linear Formula H2C=CH(CH2)9OH
Refractive Index 1.4490 to 1.4510 (20°C, 589nm)
Quantity 100 g
Beilstein 01, 452
Formula Weight 170.29
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material ω-Undecylenyl alcohol, 99%
Show More Show Less
EINECSNumber 203-971-0
RTECSNumber YQ2985000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.