Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecylphosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. U00885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00885G 5 g
U00881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00885G Supplier TCI America Supplier No. U00885G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5137-69-9
Molecular Formula C11H25O3P
Molecular Weight (g/mol) 236.29
MDL Number MFCD00015831
InChI Key GKIQHTGBORJXKZ-UHFFFAOYSA-N
PubChem CID 22112692
IUPAC Name undecylphosphonic acid
SMILES CCCCCCCCCCCP(O)(O)=O

Specifications

Melting Point 103°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 236.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Undecylphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.