Learn More
Abcam Undecylprodigiosin, Apoptotic and anticancer agent, 250UG
Catalog No. 502705825
Encompass_Preferred
MW 393.6 Da, Purity >93%. Apoptotic and anticancer agent. Selectively induces p53-independant apoptosis in cancer cells. Induces G2/M cell cycle arrest and shows antiproliferative effects. Shows antimalarial and antibactieral effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5825 Supplier Abcam Supplier No. AB144382250UG
MW 393.6 Da, Purity >93%. Apoptotic and anticancer agent. Selectively induces p53-independant apoptosis in cancer cells. Induces G2/M cell cycle arrest and shows antiproliferative effects. Shows antimalarial and antibactieral effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement