Learn More
Andwin Scientific Yamato Undercounter Ultra Low Freezer -40ºc To -85ºc, 1 Cu.Ft., Manual Defrost
SDP
Catalog No. NC1869590
Encompass_Preferred
Ultra-Low undercounter style freezer comes standard with digital controller, audible and visual alarms, remote alarm contacts, data logging, alarm battery back-up.
Catalog No. NC1869590 Supplier Andwin Scientific Supplier No. ULF001UN
Add to Cart
Add to Cart
Request a Quote
Ultra-Low undercounter style freezer comes standard with digital controller, audible and visual alarms, remote alarm contacts, data logging, alarm battery back-up.