Learn More
Yamato Scientific America, Inc. Yamato Undercounter Ultra Low Freezer -40ºc To -85ºc, 3 Cu.Ft., Manual Defrost
Catalog No. NC1869593 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Ultra-Low undercounter style freezer comes standard with digital controller, audible and visual alarms, remote alarm contacts, data logging, alarm battery back-up.
Catalog No. NC1869593 Supplier Yamato Scientific America, Inc. Supplier No. ULF201UN
Add to Cart
Add to Cart
Request a Quote
Ultra-Low undercounter style freezer comes standard with digital controller, audible and visual alarms, remote alarm contacts, data logging, alarm battery back-up.